Assay Details
Target Gene Details
Entrez Gene ID: | 151556 |
Gene Name: | G protein-coupled receptor 155 |
Gene Aliases: |
DEP.7, DEPDC3, PGR22 |
Location: |
Chr.2:174431571-174494001 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR155 | NM_001033045.3 | NP_001028217.1 | ||
| NM_001267050.1 | NP_001253979.1 | |||
| NM_001267051.1 | NP_001253980.1 | |||
| NM_152529.6 | NP_689742.4 | |||
| XM_017003485.1 | XP_016858974.1 | |||
| XM_017003486.1 | XP_016858975.1 | |||
| XM_017003487.1 | XP_016858976.1 | |||
| AK056381.1 | BAB71170.1 | |||
| AK096665.1 | BAC04835.1 | |||
| AK315125.1 | ||||
| AL834154.1 | CAH10683.1 | |||
| BC028730.1 | AAH28730.1 | |||
| BC035037.1 | AAH35037.1 | |||
| BC036681.1 | AAH36681.1 | |||
| BX537947.1 | CAD97915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583667 | Chr.2:174410951 - 174466210 on Build GRCh38 | Loss |
|
| nsv526187 | Chr.2:174421143 - 174474776 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map