Assay Details
Target Gene Details
Entrez Gene ID: | 6654 |
Gene Name: | SOS Ras/Rac guanine nucleotide exchange factor 1 |
Gene Aliases: |
GF1, GGF1, GINGF, HGF, NS4 |
Location: |
Chr.2:38981549-39124959 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOS1 | NM_005633.3 | 23 | 7284 | NP_005624.2 |
More Information
Set Membership: |
|

Genomic Map