Assay Details
Target Gene Details
Entrez Gene ID: | 1386 |
Gene Name: | activating transcription factor 2 |
Gene Aliases: |
CRE-BP1, CREB-2, CREB2, HB16, TREB7 |
Location: |
Chr.2:175072250-175168206 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF2 | NM_001256090.1 | 15 | 2825 | NP_001243019.1 |
| NM_001256091.1 | 14 | 2750 | NP_001243020.1 | |
| NM_001256092.1 | 13 | 2693 | NP_001243021.1 | |
| NM_001256093.1 | 13 | 2725 | NP_001243022.1 | |
| NM_001880.3 | 14 | 2790 | NP_001871.2 | |
| NR_045768.1 | 17 | 2987 | ||
| NR_045769.1 | 17 | 3019 | ||
| NR_045770.1 | 16 | 2872 | ||
| NR_045771.1 | 15 | 2864 | ||
| NR_045772.1 | 15 | 2802 | ||
| NR_045773.1 | 14 | 2767 | ||
| NR_045774.1 | 13 | 2675 | ||
| AF283776.2 | 1 | 411 | ||
| AK128731.1 | ||||
| AL050192.1 | 1 | 403 | ||
| BC107698.1 | 13 | 2660 | AAI07699.1 | |
| BX648469.1 | 13 | 2580 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575313 | Chr.2:174591458 - 175085724 on Build GRCh38 | Gain |
|
| nsv1012709 | Chr.2:175023400 - 175840083 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map