Assay Details
Target Gene Details
Entrez Gene ID: | 274 |
Gene Name: | bridging integrator 1 |
Gene Aliases: |
AMPH2, AMPHL, SH3P9 |
Location: |
Chr.2:127048023-127107400 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583032 | Chr.2:127013430 - 127087159 on Build GRCh38 | Loss |
|
| nsv954812 | Chr.2:127039525 - 127106424 on Build GRCh38 | Deletion |
|
| nsv516759 | Chr.2:127039090 - 127101865 on Build GRCh38 | Loss |
|
| dgv4064n100 | Chr.2:127036712 - 127104681 on Build GRCh38 | Gain |
|
| nsv999422 | Chr.2:126707071 - 127198967 on Build GRCh38 | Gain |
|
| nsv583033 | Chr.2:127043405 - 127110859 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map