Assay Details
Target Gene Details
Entrez Gene ID: | 2281 |
Gene Name: | FK506 binding protein 1B |
Gene Aliases: |
FKBP12.6, FKBP1L, OTK4, PKBP1L, PPIase |
Location: |
Chr.2:24033205-24067743 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP1B | NM_001322963.1 | NP_001309892.1 | ||
| NM_001322964.1 | NP_001309893.1 | |||
| NM_004116.4 | NP_004107.1 | |||
| NM_054033.3 | NP_473374.1 | |||
| NR_136536.1 | ||||
| NR_136538.1 | ||||
| NR_136539.1 | ||||
| XM_011532699.2 | XP_011531001.1 | |||
| XM_017003593.1 | XP_016859082.1 | |||
| XM_017003594.1 | XP_016859083.1 | |||
| XM_017003596.1 | XP_016859085.1 | |||
| AF322070.1 | AAK11191.1 | |||
| BC002614.1 | AAH02614.1 | |||
| BF216711.1 | ||||
| BF969580.1 | ||||
| BI546013.1 | ||||
| BM802489.1 | ||||
| BU147063.1 | ||||
| BU508033.1 | ||||
| D38037.1 | BAA07232.1 | |||
| L37086.1 | AAC37581.1 | |||
| S69800.1 | AAB30685.1 | |||
| S69815.1 | AAB30684.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain |
|
| nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map