Assay Details
Target Gene Details
Entrez Gene ID: | 4664 |
Gene Name: | NGFI-A binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.2:190648860-190692766 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAB1 | NM_001321312.1 | NP_001308241.1 | ||
| NM_001321313.1 | NP_001308242.1 | |||
| NM_005966.3 | NP_005957.2 | |||
| XM_005246582.1 | XP_005246639.1 | |||
| XM_005246583.1 | XP_005246640.1 | |||
| XM_011511219.2 | XP_011509521.1 | |||
| XM_017004170.1 | XP_016859659.1 | |||
| XM_017004171.1 | XP_016859660.1 | |||
| XM_017004172.1 | XP_016859661.1 | |||
| XM_017004173.1 | XP_016859662.1 | |||
| XM_017004174.1 | XP_016859663.1 | |||
| XM_017004175.1 | XP_016859664.1 | |||
| XM_017004176.1 | XP_016859665.1 | |||
| AK292446.1 | ||||
| BC035724.1 | AAH35724.1 | |||
| DB457461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3082 | Chr.2:190627513 - 190688068 on Build GRCh38 | Insertion |
|
| nsv834492 | Chr.2:190574138 - 190781895 on Build GRCh38 | Gain |
|
| dgv1200e212 | Chr.2:190638342 - 190674560 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map