Assay Details
Target Gene Details
Entrez Gene ID: | 3329 |
Gene Name: | heat shock protein family D (Hsp60) member 1 |
Gene Aliases: |
CPN60, GROEL, HLD4, HSP-60, HSP60, HSP65, HuCHA60, SPG13 |
Location: |
Chr.2:197486584-197500274 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPD1 | NM_002156.4 | NP_002147.2 | ||
| NM_199440.1 | NP_955472.1 | |||
| AK298632.1 | ||||
| AK298844.1 | ||||
| AK312240.1 | ||||
| BC002676.2 | AAH02676.1 | |||
| BC003030.1 | AAH03030.1 | |||
| BC067082.1 | AAH67082.1 | |||
| BC073746.1 | AAH73746.1 | |||
| BX647145.1 | ||||
| EU719626.1 | ||||
| KU178139.1 | ||||
| KU178140.1 | ||||
| KU178141.1 | ||||
| M22382.1 | AAA60127.1 | |||
| M34664.1 | AAA36022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014881 | Chr.2:197432432 - 197668882 on Build GRCh38 | Gain |
|
| nsv963757 | Chr.2:197486354 - 197489175 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map