Assay Details
Target Gene Details
Entrez Gene ID: | 80146 |
Gene Name: | UDP-glucuronate decarboxylase 1 |
Gene Aliases: |
SDR6E1, UGD |
Location: |
Chr.2:106093303-106194339 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
More Information
Set Membership: |
|

Genomic Map