Assay Details
Target Gene Details
Entrez Gene ID: | 80235 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class Z |
Gene Aliases: |
GPI-MT-IV, PIG-Z, SMP3 |
Location: |
Chr.3:196946343-196969411 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGZ | NM_025163.3 | NP_079439.2 | ||
| XM_011513190.1 | XP_011511492.1 | |||
| XM_011513192.2 | XP_011511494.1 | |||
| XM_017007240.1 | XP_016862729.1 | |||
| XM_017007241.1 | XP_016862730.1 | |||
| XM_017007242.1 | XP_016862731.1 | |||
| AK022830.1 | BAB14263.1 | |||
| BC044640.1 | AAH44640.1 | |||
| BG760692.1 | ||||
| DA736224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822421 | Chr.3:196949827 - 197027448 on Build GRCh38 | Loss |
|
| esv3599293 | Chr.3:196759100 - 197444213 on Build GRCh38 | Gain |
|
| nsv1003135 | Chr.3:196621850 - 197012082 on Build GRCh38 | Gain |
|
| esv2763775 | Chr.3:196727069 - 197015419 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79424507] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map