Assay Details
Target Gene Details
Entrez Gene ID: | 101060195 |
Gene Name: | FCF1 pseudogene 2 |
Gene Aliases: |
- |
Location: |
Chr.3:48290279-48291678 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCF1P2 | NR_134638.1 | 1 | 767 | |
| BU630116.1 | ||||
| CU451957.1 | 1 | 260 |
Target Gene Details
Entrez Gene ID: | 10201 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 6 |
Gene Aliases: |
IPIA-ALPHA, NDK 6, NM23-H6 |
Location: |
Chr.3:48289369-48302259 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME6 | XM_017005511.1 | XP_016861000.1 | ||
| XM_017005512.1 | 3 | 4960 | XP_016861001.1 | |
| XM_017005513.1 | 6 | 4351 | XP_016861002.1 | |
| XM_017005514.1 | 7 | 4608 | XP_016861003.1 | |
| XM_017005515.1 | 5 | 4364 | XP_016861004.1 | |
| XM_017005516.1 | XP_016861005.1 | |||
| XM_017005517.1 | 3 | 4126 | XP_016861006.1 | |
| XM_017005518.1 | 4 | 4255 | XP_016861007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834683 | Chr.3:48260014 - 48451855 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map