Assay Details
Target Gene Details
Entrez Gene ID: | 100996539 |
Gene Name: | ITPR1 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.3:4490317-4493163 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPR1-AS1 | NR_108075.1 | 2 | 752 | |
| AL832729.1 | 1 | 2179 | ||
| BI523552.1 | 2 | 780 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422450 | Chr.3:4467187 - 4614541 on Build GRCh38 | Deletion |
|

Genomic Map