Assay Details
Target Gene Details
Entrez Gene ID: | 201595 |
Gene Name: | STT3B, catalytic subunit of the oligosaccharyltransferase complex |
Gene Aliases: |
CDG1X, SIMP, STT3-B |
Location: |
Chr.3:31532501-31637622 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STT3B | NM_178862.2 | NP_849193.1 | ||
| XM_011533465.1 | XP_011531767.1 | |||
| XM_017005857.1 | XP_016861346.1 | |||
| XM_017005858.1 | XP_016861347.1 | |||
| XM_017005859.1 | XP_016861348.1 | |||
| AK074587.1 | ||||
| AK311488.1 | ||||
| AX799082.1 | CAE48447.1 | |||
| AY074880.1 | AAL71884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
|
| nsv954458 | Chr.3:31535409 - 31660708 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77681977] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map