Assay Details
Target Gene Details
Entrez Gene ID: | 105376997 |
Gene Name: | uncharacterized LOC105376997 |
Gene Aliases: |
- |
Location: |
Chr.3:25383105-25384927 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105376997 | NR_134933.1 | |||
| AK131021.1 | ||||
| AW273730.1 | ||||
| BX098796.1 |
Target Gene Details
Entrez Gene ID: | 5915 |
Gene Name: | retinoic acid receptor beta |
Gene Aliases: |
HAP, MCOPS12, NR1B2, RARbeta1, RRB2 |
Location: |
Chr.3:24829323-25597932 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RARB | NM_001290216.2 | NP_001277145.1 | ||
| NM_001290300.1 | NP_001277229.1 | |||
| AK299756.1 | ||||
| DC376623.1 | ||||
| DQ083391.1 | AAZ32403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893676 | Chr.3:24984204 - 26577498 on Build GRCh38 | Loss |
|
| nsv834645 | Chr.3:25322232 - 25513616 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78963521] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map