Assay Details
Target Gene Details
Entrez Gene ID: | 55129 |
Gene Name: | anoctamin 10 |
Gene Aliases: |
SCAR10, TMEM16K |
Location: |
Chr.3:43366324-43691594 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANO10 | NM_001204831.1 | NP_001191760.1 | ||
| NM_001204832.1 | NP_001191761.1 | |||
| NM_001204833.1 | NP_001191762.1 | |||
| NM_001204834.1 | NP_001191763.1 | |||
| NM_018075.3 | NP_060545.3 | |||
| XM_011533887.2 | XP_011532189.1 | |||
| XM_017006716.1 | XP_016862205.1 | |||
| XM_017006717.1 | XP_016862206.1 | |||
| XM_017006719.1 | XP_016862208.1 | |||
| XM_017006720.1 | XP_016862209.1 | |||
| XM_017006721.1 | XP_016862210.1 | |||
| XM_017006722.1 | XP_016862211.1 | |||
| AK096302.1 | ||||
| AK131223.1 | ||||
| AK292368.1 | ||||
| AK295969.1 | ||||
| AK297949.1 | ||||
| AL832508.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74784606] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map