Assay Details
Target Gene Details
Entrez Gene ID: | 54681 |
Gene Name: | prolyl 4-hydroxylase, transmembrane |
Gene Aliases: |
EGLN4, HIFPH4, P4H-TM, PH-4, PH4, PHD4 |
Location: |
Chr.3:48989908-49007149 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P4HTM | NM_177938.2 | NP_808807.2 | ||
| NM_177939.2 | NP_808808.1 | |||
| AK000269.1 | BAA91045.1 | |||
| AL713728.1 | CAD28518.2 | |||
| AY198406.1 | AAO43431.1 | |||
| BC011710.2 | AAH11710.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954487 | Chr.3:48993668 - 49001867 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map