Assay Details
Target Gene Details
Entrez Gene ID: | 55164 |
Gene Name: | SHQ1, H/ACA ribonucleoprotein assembly factor |
Gene Aliases: |
GRIM-1, Shq1p |
Location: |
Chr.3:72725703-72848475 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHQ1 | NM_018130.2 | NP_060600.2 | ||
| AK001401.1 | BAA91669.1 | |||
| AK092962.1 | ||||
| AK296789.1 | ||||
| BC017204.2 | AAH17204.1 | |||
| BC017274.2 | AAH17274.1 | |||
| BC025270.1 | AAH25270.1 | |||
| BC032671.1 | AAH32671.1 | |||
| BC039830.1 | AAH39830.1 | |||
| BC047879.2 | AAH47879.1 | |||
| BX641074.1 | CAE46037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763278 | Chr.3:72595871 - 73026595 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map