Assay Details
Target Gene Details
Entrez Gene ID: | 2632 |
Gene Name: | glucan (1,4-alpha-), branching enzyme 1 |
Gene Aliases: |
APBD, GBE, GSD4 |
Location: |
Chr.3:81489699-81761799 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBE1 | NM_000158.3 | NP_000149.3 | ||
| AB209731.1 | BAD92968.1 | |||
| AK125918.1 | ||||
| AK297939.1 | ||||
| AK300620.1 | ||||
| AK312245.1 | ||||
| BC012098.1 | AAH12098.1 | |||
| L07956.1 | AAA58642.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77096266] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map