Assay Details
Target Gene Details
Entrez Gene ID: | 9626 |
Gene Name: | guanylate cyclase activator 1C |
Gene Aliases: |
GCAP3 |
Location: |
Chr.3:108907644-108955195 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCA1C | NM_005459.3 | NP_005450.3 | ||
| XM_005247910.3 | XP_005247967.1 | |||
| XM_011513334.2 | XP_011511636.1 | |||
| AB593147.1 | ||||
| AF110002.1 | AAD19944.1 | |||
| AF110003.1 | AAD19945.1 | |||
| AV732101.1 | ||||
| BC103993.1 | AAI03994.1 | |||
| BC103994.1 | AAI03995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829662 | Chr.3:108821681 - 109005440 on Build GRCh38 | Gain |
|
| dgv4834n100 | Chr.3:108801003 - 108939054 on Build GRCh38 | Loss |
|
| dgv4836n100 | Chr.3:108877854 - 109296324 on Build GRCh38 | Gain |
|
| nsv829663 | Chr.3:108916545 - 109070871 on Build GRCh38 | Gain |
|
| esv3597336 | Chr.3:108891660 - 108931381 on Build GRCh38 | Gain |
|
| esv3568956 | Chr.3:108697937 - 108934542 on Build GRCh38 | Loss |
|
| esv3597329 | Chr.3:108759794 - 108933305 on Build GRCh38 | Gain |
|
| nsv522119 | Chr.3:108756961 - 109199392 on Build GRCh38 | Gain |
|
| nsv829661 | Chr.3:108751158 - 108940686 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map