Assay Details
Target Gene Details
Entrez Gene ID: | 79750 |
Gene Name: | zinc finger protein 385D |
Gene Aliases: |
ZNF659 |
Location: |
Chr.3:21418419-22372641 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF385D | NM_024697.2 | NP_078973.1 | ||
| XM_011534122.1 | XP_011532424.1 | |||
| XM_011534123.2 | XP_011532425.1 | |||
| XM_011534124.2 | XP_011532426.1 | |||
| XM_017007191.1 | XP_016862680.1 | |||
| XM_017007192.1 | XP_016862681.1 | |||
| XM_017007193.1 | XP_016862682.1 | |||
| XM_017007194.1 | XP_016862683.1 | |||
| XM_017007195.1 | XP_016862684.1 | |||
| XM_017007196.1 | XP_016862685.1 | |||
| XM_017007197.1 | XP_016862686.1 | |||
| XM_017007198.1 | XP_016862687.1 | |||
| XM_017007199.1 | XP_016862688.1 | |||
| XM_017007200.1 | XP_016862689.1 | |||
| XM_017007201.1 | XP_016862690.1 | |||
| XM_017007203.1 | XP_016862692.1 | |||
| AK026072.1 | BAB15350.1 | |||
| AK310298.1 | ||||
| BC007212.1 | AAH07212.1 |
Target Gene Details
Entrez Gene ID: | 100874216 |
Gene Name: | ZNF385D antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:21542789-21579959 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF385D-AS1 | NR_046731.1 | |||
| BG776698.1 | ||||
| CR738568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010460 | Chr.3:21475318 - 21798379 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map