Assay Details
Target Gene Details
Entrez Gene ID: | 54995 |
Gene Name: | 3-oxoacyl-ACP synthase, mitochondrial |
Gene Aliases: |
FASN2D, KASI, KS |
Location: |
Chr.3:25783661-25794534 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXSM | NM_001145391.1 | NP_001138863.1 | ||
| NM_017897.2 | NP_060367.1 | |||
| NR_026937.1 | ||||
| XM_006713216.3 | 1 | 6663 | XP_006713279.1 | |
| XM_006713217.3 | 1 | 6663 | XP_006713280.1 | |
| XM_017006713.1 | 1 | 6663 | XP_016862202.1 | |
| XM_017006714.1 | 1 | 6663 | XP_016862203.1 | |
| AK000611.1 | BAA91286.1 | |||
| AK225260.1 | ||||
| AK304821.1 | ||||
| BC008202.1 | AAH08202.1 | |||
| DA916187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893676 | Chr.3:24984204 - 26577498 on Build GRCh38 | Loss |
|
| dgv71n17 | Chr.3:25786051 - 25816479 on Build GRCh38 | Loss |
|
| esv2763742 | Chr.3:25788356 - 25868676 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map