Assay Details
Target Gene Details
Entrez Gene ID: | 64747 |
Gene Name: | major facilitator superfamily domain containing 1 |
Gene Aliases: |
SMAP4 |
Location: |
Chr.3:158801870-158829719 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD1 | NM_001167903.1 | NP_001161375.1 | ||
| NM_001289406.1 | NP_001276335.1 | |||
| NM_001289407.1 | NP_001276336.1 | |||
| NM_022736.2 | NP_073573.2 | |||
| NR_110328.1 | ||||
| XM_006713730.2 | XP_006713793.1 | |||
| AB014732.1 | BAB20269.1 | |||
| AF351617.2 | AAN76517.2 | |||
| AK024215.1 | BAB14852.1 | |||
| AK294628.1 | ||||
| AK297593.1 | ||||
| AK300497.1 | ||||
| AK301680.1 | ||||
| BC030542.1 | ||||
| BC042197.1 | AAH42197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014255 | Chr.3:158641071 - 159050753 on Build GRCh38 | Gain |
|
| nsv1006146 | Chr.3:158758035 - 158907989 on Build GRCh38 | Loss |
|
| nsv592123 | Chr.3:158760007 - 158907339 on Build GRCh38 | Loss |
|
| nsv1004124 | Chr.3:158428926 - 159090849 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74951656] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map