Assay Details
Target Gene Details
Entrez Gene ID: | 27032 |
Gene Name: | ATPase secretory pathway Ca2+ transporting 1 |
Gene Aliases: |
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1 |
Location: |
Chr.3:130850500-131016712 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005160 | Chr.3:130723453 - 131010147 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
