Assay Details
Target Gene Details
Entrez Gene ID: | 100874101 |
Gene Name: | HDAC11 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:13476987-13480053 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC11-AS1 | NR_046690.1 | |||
| BX104155.1 |

Genomic Map