Assay Details
Target Gene Details
Entrez Gene ID: | 101929130 |
Gene Name: | uncharacterized LOC101929130 |
Gene Aliases: |
- |
Location: |
Chr.3:187291272-187297933 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929130 | NR_135551.1 | |||
| BX648496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009271 | Chr.3:187246858 - 187480081 on Build GRCh38 | Gain |
|
| nsv1014596 | Chr.3:187106902 - 187410668 on Build GRCh38 | Gain |
|

Genomic Map