Assay Details
Target Gene Details
Entrez Gene ID: | 10425 |
Gene Name: | ariadne RBR E3 ubiquitin protein ligase 2 |
Gene Aliases: |
ARI2, TRIAD1 |
Location: |
Chr.3:48918820-48985541 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2473053 | Chr.3:48919846 - 48922072 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map