Assay Details
Target Gene Details
Entrez Gene ID: | 10954 |
Gene Name: | protein disulfide isomerase family A member 5 |
Gene Aliases: |
PDIR |
Location: |
Chr.3:123067009-123162106 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDIA5 | NM_006810.3 | NP_006801.1 | ||
| NR_028444.1 | ||||
| BM549365.1 |
More Information
Set Membership: |
|

Genomic Map