Assay Details
Target Gene Details
Entrez Gene ID: | 26137 |
Gene Name: | zinc finger and BTB domain containing 20 |
Gene Aliases: |
DPZF, HOF, ODA-8S, PRIMS, ZNF288 |
Location: |
Chr.3:114314500-115147280 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB20 | NM_001164342.2 | NP_001157814.1 | ||
| NM_001164343.2 | NP_001157815.1 | |||
| NM_001164344.2 | NP_001157816.1 | |||
| NM_001164345.2 | NP_001157817.1 | |||
| NM_001164346.2 | NP_001157818.1 | |||
| NM_001164347.2 | NP_001157819.1 | |||
| NM_015642.5 | NP_056457.3 | |||
| NR_121662.1 | ||||
| AF139460.2 | AAG28340.2 | |||
| AK225788.1 | ||||
| AK315307.1 | ||||
| AL050276.1 | CAB43377.1 | |||
| BC029041.1 | AAH29041.1 | |||
| BG993862.1 | ||||
| BX647778.1 | CAH56190.1 | |||
| CB164748.1 | ||||
| DB268190.1 |
Target Gene Details
Entrez Gene ID: | 100131117 |
Gene Name: | ZBTB20 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:114351811-114388978 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB20-AS1 | NR_038993.1 | |||
| AK098660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829679 | Chr.3:114331081 - 114533888 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114954555] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map