Assay Details
Target Gene Details
Entrez Gene ID: | 116832 |
Gene Name: | ribosomal protein L39 like |
Gene Aliases: |
RPL39L1 |
Location: |
Chr.3:187120953-187139475 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL39L | NM_052969.1 | NP_443201.1 | ||
| AB063610.1 | BAC19837.1 | |||
| BC012328.1 | AAH12328.1 | |||
| L05096.1 | AAC15859.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004008 | Chr.3:186879814 - 187204053 on Build GRCh38 | Gain |
|
| nsv1014596 | Chr.3:187106902 - 187410668 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map