Assay Details
Target Gene Details
Entrez Gene ID: | 10201 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 6 |
Gene Aliases: |
IPIA-ALPHA, NDK 6, NM23-H6 |
Location: |
Chr.3:48289369-48302259 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME6 | NM_001308426.1 | 6 | 2153 | NP_001295355.1 |
| NM_001308427.1 | 6 | 1962 | NP_001295356.1 | |
| NM_001308428.1 | 6 | 1953 | NP_001295357.1 | |
| NM_001308430.1 | 5 | 1896 | NP_001295359.1 | |
| NM_001308431.1 | 5 | 2113 | NP_001295360.1 | |
| NM_001308433.1 | 5 | 1913 | NP_001295362.1 | |
| NM_001308434.1 | 5 | 2056 | NP_001295363.1 | |
| NM_001308435.1 | 4 | 1952 | NP_001295364.1 | |
| NM_005793.4 | 6 | 1936 | NP_005784.1 | |
| XM_017005511.1 | XP_016861000.1 | |||
| XM_017005512.1 | 3 | 2487 | XP_016861001.1 | |
| XM_017005513.1 | 6 | 1878 | XP_016861002.1 | |
| XM_017005514.1 | 7 | 2135 | XP_016861003.1 | |
| XM_017005515.1 | 5 | 1891 | XP_016861004.1 | |
| XM_017005516.1 | XP_016861005.1 | |||
| XM_017005517.1 | 3 | 1653 | XP_016861006.1 | |
| XM_017005518.1 | 4 | 1782 | XP_016861007.1 | |
| CN287719.1 | 1 | 517 | ||
| DA519225.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834683 | Chr.3:48260014 - 48451855 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map