Assay Details
Target Gene Details
Entrez Gene ID: | 10098 |
Gene Name: | tetraspanin 5 |
Gene Aliases: |
NET-4, NET4, TM4SF9, TSPAN-5 |
Location: |
Chr.4:98470367-98658661 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN5 | NM_005723.3 | NP_005714.2 | ||
| XM_005262680.1 | XP_005262737.1 | |||
| AF065389.1 | AAC17120.1 | |||
| AK295385.1 | ||||
| AK315723.1 | ||||
| BC009704.1 | AAH09704.1 | |||
| CR541809.1 | CAG46608.1 | |||
| CR541829.1 | CAG46628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004181 | Chr.4:98287001 - 98608425 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map