Assay Details
Target Gene Details
Entrez Gene ID: | 80155 |
Gene Name: | N(alpha)-acetyltransferase 15, NatA auxiliary subunit |
Gene Aliases: |
Ga19, NARG1, NAT1P, NATH, TBDN, TBDN100 |
Location: |
Chr.4:139301467-139390781 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA15 | NM_057175.3 | NP_476516.1 | ||
| XM_005263236.2 | XP_005263293.1 | |||
| AF327722.1 | AAK15707.1 | |||
| AJ314788.1 | CAC43228.1 | |||
| AK023387.1 | ||||
| AK023402.1 | BAB14562.1 | |||
| AK304417.1 | ||||
| AK314626.1 | ||||
| AY039242.1 | AAK68661.1 | |||
| AY112670.1 | AAM48746.1 | |||
| BC032642.1 | AAH32642.1 | |||
| BC039818.1 | AAH39818.1 | |||
| BC093928.1 | AAH93928.1 | |||
| BC104806.1 | AAI04807.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034429 | Chr.4:139302827 - 139343882 on Build GRCh38 | Loss |
|
| dgv9292n54 | Chr.4:139307207 - 139317207 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map