Assay Details
Target Gene Details
Entrez Gene ID: | 80157 |
Gene Name: | cell wall biogenesis 43 C-terminal homolog |
Gene Aliases: |
CWH43-C |
Location: |
Chr.4:48986247-49062079 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CWH43 | NM_001286791.1 | NP_001273720.1 | ||
| NM_025087.2 | NP_079363.2 | |||
| XM_011513755.1 | XP_011512057.1 | |||
| XM_011513756.2 | XP_011512058.1 | |||
| XM_011513757.2 | XP_011512059.1 | |||
| XM_011513758.1 | XP_011512060.1 | |||
| AK025164.1 | BAB15080.1 | |||
| AK300495.1 | ||||
| AK316307.1 | ||||
| BC137387.1 | ||||
| BC137388.1 | ||||
| DA426878.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010381 | Chr.4:48699508 - 49307825 on Build GRCh38 | Gain |
|
| nsv1161082 | Chr.4:48976224 - 48998069 on Build GRCh38 | Duplication |
|
| nsv1073757 | Chr.4:48974954 - 48999283 on Build GRCh38 | Deletion |
|
| esv3893921 | Chr.4:48934804 - 49089765 on Build GRCh38 | Gain |
|
| esv3600473 | Chr.4:48978121 - 48993691 on Build GRCh38 | Loss |
|
| nsv441875 | Chr.4:48977617 - 48991360 on Build GRCh38 | Loss |
|
| nsv1011910 | Chr.4:48782315 - 49152740 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map