Assay Details
Target Gene Details
Entrez Gene ID: | 2868 |
Gene Name: | G protein-coupled receptor kinase 4 |
Gene Aliases: |
GPRK2L, GPRK4, GRK4a, IT11 |
Location: |
Chr.4:2963504-3040760 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575673 | Chr.4:2957494 - 3022996 on Build GRCh38 | Gain |
|
| esv3599416 | Chr.4:2972522 - 3012111 on Build GRCh38 | Gain |
|
| nsv593438 | Chr.4:2946729 - 3028370 on Build GRCh38 | Gain |
|
| dgv5082n100 | Chr.4:2956087 - 3017315 on Build GRCh38 | Gain |
|
| nsv998311 | Chr.4:2979888 - 3010996 on Build GRCh38 | Loss |
|
| esv3599415 | Chr.4:2958013 - 3019896 on Build GRCh38 | Gain |
|
| esv3599412 | Chr.4:2940424 - 3030117 on Build GRCh38 | Gain |
|
| nsv461163 | Chr.4:2904558 - 3233253 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map