Assay Details
Target Gene Details
Entrez Gene ID: | 7468 |
Gene Name: | Wolf-Hirschhorn syndrome candidate 1 |
Gene Aliases: |
MMSET, NSD2, REIIBP, TRX5, WHS |
Location: |
Chr.4:1871357-1982207 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WHSC1 | NM_001042424.2 | NP_001035889.1 | ||
| NM_007331.1 | NP_015627.1 | |||
| NM_133330.2 | NP_579877.1 | |||
| NM_133331.2 | NP_579878.1 | |||
| NM_133335.3 | NP_579890.1 | |||
| XM_005248002.2 | XP_005248059.1 | |||
| XM_005248005.2 | XP_005248062.1 | |||
| XM_011513557.2 | XP_011511859.1 | |||
| AF083386.1 | AAD19343.1 | |||
| AF083387.1 | AAD21770.1 | |||
| AF083388.1 | AAD21771.1 | |||
| AF083389.1 | AAD19344.1 | |||
| AF083390.1 | AAD19345.1 | |||
| AF083391.1 | AAD19346.1 | |||
| AJ007042.1 | CAB45386.1 | |||
| AK289697.1 | ||||
| BC020545.1 | ||||
| BC052254.1 | AAH52254.1 | |||
| CX866583.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829836 | Chr.4:1775678 - 1901525 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map