Assay Details
Target Gene Details
Entrez Gene ID: | 4717 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit C1 |
Gene Aliases: |
KFYI |
Location: |
Chr.4:139289917-139302551 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFC1 | NM_001184986.1 | NP_001171915.1 | ||
| NM_001184987.1 | NP_001171916.1 | |||
| NM_001184988.1 | NP_001171917.1 | |||
| NM_001184989.1 | NP_001171918.1 | |||
| NM_001184990.1 | NP_001171919.1 | |||
| NM_001184991.1 | NP_001171920.1 | |||
| NM_002494.3 | NP_002485.1 | |||
| AF047184.1 | AAC04269.1 | |||
| AF047435.1 | AAC39886.1 | |||
| AK291147.1 | ||||
| BC101798.1 | AAI01799.1 | |||
| BC107682.1 | AAI07683.1 | |||
| BC112061.1 | AAI12062.1 | |||
| BG701467.1 | ||||
| BM768825.1 | ||||
| BU677336.1 | ||||
| CA313899.1 | ||||
| CB047240.1 | ||||
| CB160001.1 | ||||
| CN347371.1 | ||||
| CR457005.1 | CAG33286.1 | |||
| W56292.1 |
More Information
Set Membership: |
|

Genomic Map