Assay Details
Target Gene Details
Entrez Gene ID: | 7424 |
Gene Name: | vascular endothelial growth factor C |
Gene Aliases: |
Flt4-L, LMPH1D, VRP |
Location: |
Chr.4:176683534-176792745 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VEGFC | NM_005429.4 | NP_005420.1 | ||
| AK313879.1 | ||||
| BC035212.1 | AAH35212.1 | |||
| BC063685.1 | AAH63685.1 | |||
| CR541897.1 | CAG46695.1 | |||
| U43142.1 | AAA85214.1 | |||
| U58111.1 | AAB02909.1 | |||
| X94216.1 | CAA63907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2728772 | Chr.4:175817087 - 177875422 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74657378] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map