Assay Details
Target Gene Details
Entrez Gene ID: | 57587 |
Gene Name: | cilia and flagella associated protein 97 |
Gene Aliases: |
KIAA1430, hmw |
Location: |
Chr.4:185159662-185210251 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP97 | XM_017008485.1 | XP_016863974.1 |
Target Gene Details
Entrez Gene ID: | 83891 |
Gene Name: | sorting nexin 25 |
Gene Aliases: |
MSTP043, SBBI31 |
Location: |
Chr.4:185204237-185363971 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNX25 | NM_001317781.1 | NP_001304710.1 | ||
| AY601647.1 | AAT98627.1 | |||
| BM554003.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461859 | Chr.4:184907523 - 185314394 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72706066] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map