Assay Details
Target Gene Details
Entrez Gene ID: | 51778 |
Gene Name: | myozenin 2 |
Gene Aliases: |
C4orf5, CMH16, CS-1 |
Location: |
Chr.4:119135784-119187789 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOZ2 | NM_016599.4 | NP_057683.1 | ||
| XM_006714234.3 | XP_006714297.1 | |||
| AJ252149.1 | CAB92965.1 | |||
| AK313856.1 | ||||
| AY013295.1 | AAG38938.1 | |||
| BC005195.1 | AAH05195.1 | |||
| BC017402.1 | AAH17402.1 | |||
| BC020932.1 | AAH20932.1 | |||
| BP370492.1 | ||||
| BT007214.1 | AAP35878.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3601959 | Chr.4:119060737 - 119205365 on Build GRCh38 | Gain |
|
| dgv5373n100 | Chr.4:119086758 - 119222694 on Build GRCh38 | Gain |
|
| nsv522386 | Chr.4:119087992 - 119194224 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map