Assay Details
Target Gene Details
Entrez Gene ID: | 2983 |
Gene Name: | guanylate cyclase 1 soluble subunit beta |
Gene Aliases: |
GC-S-beta-1, GC-SB3, GUC1B3, GUCB3, GUCSB3, GUCY1B1 |
Location: |
Chr.4:155758973-155807642 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCY1B3 | NM_000857.3 | NP_000848.1 | ||
| NM_001291951.1 | NP_001278880.1 | |||
| NM_001291952.1 | NP_001278881.1 | |||
| NM_001291953.1 | NP_001278882.1 | |||
| NM_001291954.1 | NP_001278883.1 | |||
| NM_001291955.1 | NP_001278884.1 | |||
| XM_011531901.1 | XP_011530203.1 | |||
| XM_017008130.1 | XP_016863619.1 | |||
| XM_017008131.1 | XP_016863620.1 | |||
| XM_017008132.1 | XP_016863621.1 | |||
| XM_017008133.1 | XP_016863622.1 | |||
| AF020340.1 | AAB94877.1 | |||
| AK296680.1 | ||||
| AK299916.1 | ||||
| AK300296.1 | ||||
| AK307838.1 | ||||
| AK315199.1 | ||||
| AK315944.1 | ||||
| BC047620.1 | AAH47620.2 | |||
| DC317249.1 | ||||
| DC317683.1 | ||||
| DC342619.1 | ||||
| X66533.1 | CAA47144.1 |
More Information
Set Membership: |
|

Genomic Map