Assay Details
Target Gene Details
Entrez Gene ID: | 439933 |
Gene Name: | uncharacterized LOC439933 |
Gene Aliases: |
- |
Location: |
Chr.4:36244116-36274215 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC439933 | NR_122079.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3600217 | Chr.4:36111290 - 36503831 on Build GRCh38 | Loss |
|
| nsv829907 | Chr.4:36158672 - 36317960 on Build GRCh38 | Gain |
|
| esv3569373 | Chr.4:35669991 - 36407345 on Build GRCh38 | Loss |
|

Genomic Map