Assay Details
Target Gene Details
Entrez Gene ID: | 1665 |
Gene Name: | DEAH-box helicase 15 |
Gene Aliases: |
DBP1, DDX15, HRH2, PRP43, PRPF43, PrPp43p |
Location: |
Chr.4:24527464-24584603 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX15 | NM_001358.2 | NP_001349.2 | ||
| AB001636.1 | BAA23987.1 | |||
| AF279891.1 | AAF90182.1 | |||
| AK055898.1 | ||||
| AK056726.1 | ||||
| AK303506.1 | ||||
| BC035974.1 | AAH35974.1 |
More Information
Set Membership: |
|

Genomic Map