Assay Details
Target Gene Details
Entrez Gene ID: | 64854 |
Gene Name: | ubiquitin specific peptidase 46 |
Gene Aliases: |
- |
Location: |
Chr.4:52590960-52659335 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP46 | NM_001134223.1 | NP_001127695.1 | ||
| NM_001286767.1 | NP_001273696.1 | |||
| NM_001286768.1 | NP_001273697.1 | |||
| NM_022832.3 | NP_073743.2 | |||
| XM_017008557.1 | XP_016864046.1 | |||
| AK022614.1 | BAB14133.1 | |||
| AK024318.1 | BAB14881.1 | |||
| AK296493.1 | ||||
| AK299883.1 | ||||
| AK302438.1 | ||||
| BC037574.1 | AAH37574.1 | |||
| GU455414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524494 | Chr.4:52593454 - 52715898 on Build GRCh38 | Loss |
|
| nsv594201 | Chr.4:51932458 - 52866278 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map