Assay Details
Target Gene Details
Entrez Gene ID: | 23284 |
Gene Name: | adhesion G protein-coupled receptor L3 |
Gene Aliases: |
CIRL3, CL3, LEC3, LPHN3 |
Location: |
Chr.4:61201256-62078335 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRL3 | NM_001322246.1 | 1 | 276 | NP_001309175.1 |
| NM_001322402.1 | 1 | 276 | NP_001309331.1 | |
| NM_015236.5 | 1 | 276 | NP_056051.2 | |
| AK309793.1 | 1 | 279 |
More Information
Set Membership: |
|

Genomic Map