Assay Details
Target Gene Details
Entrez Gene ID: | 64116 |
Gene Name: | solute carrier family 39 member 8 |
Gene Aliases: |
BIGM103, CDG2N, LZT-Hs6, PP3105, ZIP8 |
Location: |
Chr.4:102251041-102345498 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A8 | NM_001135147.1 | NP_001128619.1 | ||
| XM_017008542.1 | XP_016864031.1 | |||
| AK304274.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5348n100 | Chr.4:102253039 - 102285398 on Build GRCh38 | Gain |
|
| nsv428447 | Chr.4:102182573 - 102340950 on Build GRCh38 | Gain |
|
| dgv5346n100 | Chr.4:102232552 - 102285398 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map