Assay Details
Target Gene Details
Entrez Gene ID: | 27071 |
Gene Name: | dual adaptor of phosphotyrosine and 3-phosphoinositides 1 |
Gene Aliases: |
BAM32 |
Location: |
Chr.4:99816824-99872290 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAPP1 | NM_001306151.1 | NP_001293080.1 | ||
| NM_014395.2 | NP_055210.2 | |||
| XM_011531840.2 | XP_011530142.1 | |||
| XM_011531842.1 | XP_011530144.1 | |||
| XM_017008023.1 | XP_016863512.1 | |||
| XM_017008024.1 | XP_016863513.1 | |||
| AF161551.1 | AAF29038.1 | |||
| AF163254.1 | AAD49697.1 | |||
| AF178987.1 | AAF44351.1 | |||
| AF186022.1 | AAF14578.1 | |||
| AK301355.1 | ||||
| BC012924.1 | AAH12924.1 | |||
| DN995752.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78266626] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map