Assay Details
Target Gene Details
Entrez Gene ID: | 60558 |
Gene Name: | GUF1 homolog, GTPase |
Gene Aliases: |
EF-4 |
Location: |
Chr.4:44678395-44700685 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUF1 | NM_021927.2 | 17 | 3524 | NP_068746.2 |
| XM_005248122.3 | 16 | 3378 | XP_005248179.1 | |
| XM_011513732.2 | 16 | 3425 | XP_011512034.1 | |
| AK001607.1 | 3 | 1597 | ||
| AK025248.1 | 9 | 2304 | BAB15090.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013488 | Chr.4:44541791 - 44974106 on Build GRCh38 | Loss |
|
| nsv4319 | Chr.4:44671518 - 44716896 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map