Assay Details
Target Gene Details
Entrez Gene ID: | 79682 |
Gene Name: | centromere protein U |
Gene Aliases: |
CENP50, CENPU50, KLIP1, MLF1IP, PBIP1 |
Location: |
Chr.4:184694065-184734132 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPU | NM_024629.3 | NP_078905.2 | ||
| NR_104593.1 | ||||
| XM_005263218.4 | XP_005263275.2 | |||
| AF469667.1 | AAQ05290.1 | |||
| AF516710.1 | AAQ08228.1 | |||
| AK027121.1 | BAB15665.1 | |||
| AK292297.1 | ||||
| BC031520.1 | AAH31520.1 | |||
| BC107744.1 | AAI07745.1 | |||
| BC131556.1 | ||||
| BC141854.1 | ||||
| CR457376.1 | CAG33657.1 | |||
| DQ907910.1 | ||||
| DQ907911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830168 | Chr.4:184626297 - 184801578 on Build GRCh38 | Loss |
|
| nsv1026074 | Chr.4:184384963 - 184780663 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map