Assay Details
Target Gene Details
Entrez Gene ID: | 419 |
Gene Name: | ADP-ribosyltransferase 3 |
Gene Aliases: |
ARTC3 |
Location: |
Chr.4:76011180-76112802 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ART3 | NM_001130017.2 | NP_001123489.1 | ||
| AK225860.1 | ||||
| AY543656.1 | AAT10278.1 | |||
| AY543657.1 | AAT10279.1 | |||
| AY543658.1 | AAT10280.1 | |||
| AY543659.1 | AAT10281.1 | |||
| DB451386.1 | ||||
| U47054.1 | AAB01894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv27112 | Chr.4:76027416 - 76056627 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map