Assay Details
Target Gene Details
Entrez Gene ID: | 5431 |
Gene Name: | RNA polymerase II subunit B |
Gene Aliases: |
POL2RB, RPB2, hRPB140 |
Location: |
Chr.4:56978640-57031162 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2B | NM_000938.2 | NP_000929.1 | ||
| NM_001303268.1 | NP_001290197.1 | |||
| NM_001303269.1 | NP_001290198.1 | |||
| AK289823.1 | ||||
| AK294901.1 | ||||
| AK295132.1 | ||||
| AK299090.1 | ||||
| BC023503.2 | AAH23503.2 | |||
| X63563.1 | CAA45124.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5214n100 | Chr.4:56786612 - 56999637 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map