Assay Details
Target Gene Details
Entrez Gene ID: | 132789 |
Gene Name: | glucosamine-6-phosphate deaminase 2 |
Gene Aliases: |
GNP2, SB52 |
Location: |
Chr.4:44701795-44726634 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNPDA2 | NM_001270880.1 | NP_001257809.1 | ||
| NM_001270881.1 | NP_001257810.1 | |||
| NM_138335.2 | NP_612208.1 | |||
| NR_073094.1 | ||||
| NR_073095.1 | ||||
| AB209904.1 | BAD93141.1 | |||
| AF247786.1 | AAL95691.1 | |||
| AK055639.1 | BAB70977.1 | |||
| AK296051.1 | ||||
| AK308920.1 | ||||
| AL834506.1 | CAD39163.1 | |||
| AY173948.1 | AAO49718.1 | |||
| BC015532.2 | AAH15532.1 | |||
| BP274548.1 | ||||
| HY163015.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013488 | Chr.4:44541791 - 44974106 on Build GRCh38 | Loss |
|
| nsv4319 | Chr.4:44671518 - 44716896 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map